Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOSTDC1
Entrez Gene ID
523184
Gene Name
sclerostin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 24961800 - 24966045 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046265.1 NP_001039730.1 1 - 2 314 65
GenBank mRNA BC113351.1 - 1 - 2 314 65

View Full Product Details