Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2D2
Entrez Gene ID
541003
Gene Name
ubiquitin conjugating enzyme E2 D2
Gene Aliases
-
Chromosome Location
Chr.7: 50789407 - 50832731 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046496.1 NP_001039961.1 5 - 6 782 101
GenBank mRNA BC114654.1 - 5 - 6 782 101

View Full Product Details