Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EAF2
Entrez Gene ID
613523
Gene Name
ELL associated factor 2
Gene Aliases
-
Chromosome Location
Chr.1: 66294136 - 66349800 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046507.2 NP_001039972.1 3 - 4 386 79
GenBank mRNA BC112586.1 - 3 - 4 387 79

View Full Product Details