Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP300
Entrez Gene ID
617430
Gene Name
cilia and flagella associated protein 300
Gene Aliases
C11orf70, C15H11orf70, C2H11orf70
Chromosome Location
Chr.15: 6664994 - 6692983 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046598.2 NP_001040063.1 4 - 5 454 90
GenBank mRNA BC105371.1 - 4 - 5 455 90

View Full Product Details