Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP5
Entrez Gene ID
617543
Gene Name
receptor accessory protein 5
Gene Aliases
-
Chromosome Location
Chr.10: 1094713 - 1145331 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046605.1 NP_001040070.1 2 - 3 282 88
GenBank mRNA BC114113.1 - 2 - 3 282 88

View Full Product Details