Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAS2R10
Entrez Gene ID
664636
Gene Name
taste receptor, type 2, member 10
Gene Aliases
BOTA-T2R10A
Chromosome Location
Chr.5: 98749622 - 98750521 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046626.1 NP_001040091.1 1 - 1 445 128

View Full Product Details