Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGME1
Entrez Gene ID
506205
Gene Name
mitochondrial genome maintenance exonuclease 1
Gene Aliases
C13H20orf72
Chromosome Location
Chr.13: 38240341 - 38250803 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075245.1 NP_001068713.1 3 - 4 939 63
GenBank mRNA BC116086.1 - 3 - 4 939 63

View Full Product Details