Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H8
Entrez Gene ID
506983
Gene Name
zinc finger CCCH-type containing 8
Gene Aliases
-
Chromosome Location
Chr.11: 203489 - 231076 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075291.2 NP_001068759.1 7 - 8 952 110
GenBank mRNA BC118120.1 - 7 - 8 953 110

View Full Product Details