Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFDN6
Entrez Gene ID
509914
Gene Name
prefoldin subunit 6
Gene Aliases
-
Chromosome Location
Chr.23: 7463411 - 7464809 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075421.2 NP_001068889.1 3 - 4 487 78
GenBank mRNA BC118486.1 - 3 - 4 488 78

View Full Product Details