Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35G2
Entrez Gene ID
511543
Gene Name
solute carrier family 35 member G2
Gene Aliases
TMEM22
Chromosome Location
Chr.1: 132298306 - 132334597 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075524.2 NP_001068992.1 1 - 2 139 150
GenBank mRNA BC118384.1 - 1 - 2 140 150

View Full Product Details