Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNIP3
Entrez Gene ID
513316
Gene Name
potassium voltage-gated channel interacting protein 3
Gene Aliases
KChIP3
Chromosome Location
Chr.11: 2042314 - 2136545 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075612.2 NP_001069080.1 1 - 2 87 101
GenBank mRNA BC118417.1 - 1 - 2 88 101

View Full Product Details