Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFIT5
Entrez Gene ID
515091
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Gene Aliases
-
Chromosome Location
Chr.26: 11081800 - 11091762 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075698.1 NP_001069166.1 1 - 2 177 130
GenBank mRNA BC118096.1 - 1 - 2 177 130

View Full Product Details