Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRAK4
Entrez Gene ID
533692
Gene Name
interleukin 1 receptor associated kinase 4
Gene Aliases
IRAK-4
Chromosome Location
Chr.5: 36680671 - 36708802 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075998.1 NP_001069466.1 10 - 11 1258 150
GenBank mRNA BC114720.1 - 10 - 11 1258 150
EU528625.1 - 10 - 11 1434 150
EU528626.1 - 10 - 11 1276 150

View Full Product Details