Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFDP1
Entrez Gene ID
534579
Gene Name
transcription factor Dp-1
Gene Aliases
-
Chromosome Location
Chr.12: 86721172 - 86738847 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076029.1 NP_001069497.1 9 - 10 954 84
GenBank mRNA BC118103.1 - 9 - 10 954 84

View Full Product Details