The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076078.2 | NP_001069546.1 | 2 - 3 | 217 | 67 |
GenBank mRNA | BC118437.1 | - | 2 - 3 | 218 | 67 |