Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A13
Entrez Gene ID
538855
Gene Name
solute carrier family 16 member 13
Gene Aliases
MCT13
Chromosome Location
Chr.19: 26821869 - 26825361 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076132.1 NP_001069600.1 2 - 3 445 63
GenBank mRNA AB435508.1 - 2 - 3 296 63
BC118219.1 - 2 - 3 445 63

View Full Product Details