Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPA
Entrez Gene ID
613653
Gene Name
inosine triphosphatase
Gene Aliases
-
Chromosome Location
Chr.13: 51987992 - 52001220 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076282.2 NP_001069750.1 6 - 7 424 101
GenBank mRNA BC112688.1 - 6 - 7 425 101

View Full Product Details