Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSU72
Entrez Gene ID
614837
Gene Name
SSU72 homolog, RNA polymerase II CTD phosphatase
Gene Aliases
-
Chromosome Location
Chr.16: 51053560 - 51078058 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076349.1 NP_001069817.1 2 - 3 301 128
GenBank mRNA BC118344.1 - 2 - 3 301 128

View Full Product Details