Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAUS2
Entrez Gene ID
508698
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
CEP27
Chromosome Location
Chr.10: 37875094 - 37888395 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076819.2 NP_001070287.1 3 - 4 263 127
GenBank mRNA BC105449.1 - 3 - 4 268 127
BT030707.1 - 3 - 4 263 127

View Full Product Details