Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABHD8
Entrez Gene ID
768306
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
Chromosome Location
Chr.7: 5822902 - 5829859 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078123.1 NP_001071591.1 3 - 4 990 67
GenBank mRNA BC118251.1 - 3 - 4 990 67

View Full Product Details