The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078129.2 | NP_001071597.1 | 5 - 6 | 411 | 73 |
GenBank mRNA | BC105388.1 | - | 5 - 6 | 412 | 73 |