Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPATCH4
Entrez Gene ID
768314
Gene Name
G-patch domain containing 4
Gene Aliases
GPATC4
Chromosome Location
Chr.3: 14232874 - 14240974 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078129.2 NP_001071597.1 7 - 8 598 78
GenBank mRNA BC105388.1 - 7 - 8 599 78

View Full Product Details