Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRA1
Entrez Gene ID
780787
Gene Name
steroid receptor RNA activator 1
Gene Aliases
-
Chromosome Location
Chr.7: 51724907 - 51731930 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079642.2 NP_001073110.1 3 - 4 378 75
GenBank mRNA BC105460.1 - 3 - 4 379 75

View Full Product Details