Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGPAT2
Entrez Gene ID
512112
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.11: 104087838 - 104100209 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080264.1 NP_001073733.1 5 - 6 768 138
GenBank mRNA BC114681.1 - 5 - 6 768 138

View Full Product Details