Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TUT1
Entrez Gene ID
616339
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
RBM21
Chromosome Location
Chr.29: 40927674 - 40937085 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080322.1 NP_001073791.1 3 - 4 620 67
GenBank mRNA BT025417.1 - 3 - 4 620 67

View Full Product Details