Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK1B
Entrez Gene ID
282113
Gene Name
uroplakin 1B
Gene Aliases
-
Chromosome Location
Chr.1: 63981479 - 64010646 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174482.2 NP_776907.2 3 - 4 393 91
GenBank mRNA BC116014.1 - 3 - 4 393 91
Z29378.1 - - 325 91

View Full Product Details