Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
G6PC3
Entrez Gene ID
369023
Gene Name
glucose-6-phosphatase catalytic subunit 3
Gene Aliases
-
Chromosome Location
Chr.19: 43907069 - 43911994 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183364.3 NP_899208.2 5 - 6 713 74
GenBank mRNA AY279358.1 - 5 - 6 729 74
BC118357.1 - 5 - 6 714 74

View Full Product Details