Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A38
Entrez Gene ID
512325
Gene Name
solute carrier family 25 member 38
Gene Aliases
-
Chromosome Location
Chr.22: 12645740 - 12660768 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014905.2 NP_001014905.2 3 - 4 519 59
GenBank mRNA BC148053.1 - 3 - 4 543 59
BT020649.1 - 3 - 4 558 59
BT020652.1 - 3 - 4 552 59
BT020813.1 - 3 - 4 532 59
BT020880.1 - 3 - 4 546 59
BT021801.1 - 3 - 4 517 59
BT026157.1 - 3 - 4 519 59

View Full Product Details