Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCAT
Entrez Gene ID
319141
Gene Name
glycine C-acetyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 109622546 - 109629294 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075134.1 NP_001068602.1 8 - 9 1112 70
GenBank mRNA BC119870.1 - 8 - 9 1112 70

View Full Product Details