Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMO2
Entrez Gene ID
504401
Gene Name
flavin containing dimethylaniline monoxygenase 2
Gene Aliases
-
Chromosome Location
Chr.16: 38756046 - 38800243 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075162.2 NP_001068630.1 3 - 4 444 124
NM_001163274.3 NP_001156746.2 3 - 4 444 124
GenBank mRNA BC120448.1 - 3 - 4 364 124
BE751148.1 - 3 - 4 444 124
EH159834.1 - 3 - 4 410 124

View Full Product Details