Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMO2
Entrez Gene ID
504401
Gene Name
flavin containing dimethylaniline monoxygenase 2
Gene Aliases
-
Chromosome Location
Chr.16: 38756046 - 38800243 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075162.2 NP_001068630.1 6 - 7 948 131
NM_001163274.3 NP_001156746.2 6 - 7 948 131
GenBank mRNA BC120448.1 - 6 - 7 868 131

View Full Product Details