Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPM6A
Entrez Gene ID
507389
Gene Name
glycoprotein M6A
Gene Aliases
-
Chromosome Location
Chr.27: 7348658 - 7606530 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075309.1 NP_001068777.1 6 - 7 1043 100
GenBank mRNA BC119902.1 - 6 - 7 1043 100

View Full Product Details