Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USB1
Entrez Gene ID
510934
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
Gene Aliases
C16orf57, C18H16orf57
Chromosome Location
Chr.18: 25907242 - 25923158 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075477.2 NP_001068945.1 4 - 5 559 134
GenBank mRNA BC122807.1 - 4 - 5 560 134

View Full Product Details