Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SURF6
Entrez Gene ID
511610
Gene Name
surfeit 6
Gene Aliases
-
Chromosome Location
Chr.11: 104240168 - 104246931 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075529.1 NP_001068997.1 1 - 2 215 88
GenBank mRNA BC119935.1 - 1 - 2 215 88

View Full Product Details