Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB9B
Entrez Gene ID
511866
Gene Name
RAB9B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.X: 52595792 - 52605804 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075541.2 NP_001069009.1 2 - 3 232 109
GenBank mRNA BC120255.1 - 2 - 3 233 109

View Full Product Details