Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAXBP1
Entrez Gene ID
514918
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
GCFC, GCFC1
Chromosome Location
Chr.1: 2786812 - 2819637 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC120358.1 - 2 - 3 468 75

View Full Product Details