Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
515608
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
-
Chromosome Location
Chr.21: 21091875 - 21105958 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075721.1 NP_001069189.1 2 - 3 288 72
GenBank mRNA BC120076.1 - 2 - 3 288 72

View Full Product Details