Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TP53I11
Entrez Gene ID
515682
Gene Name
tumor protein p53 inducible protein 11
Gene Aliases
-
Chromosome Location
Chr.15: 75004237 - 75021831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075725.1 NP_001069193.1 4 - 5 436 60
GenBank mRNA BC120050.1 - 4 - 5 436 60

View Full Product Details