Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B2
Entrez Gene ID
515710
Gene Name
hydroxysteroid 17-beta dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.18: 8586156 - 8679914 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075726.1 NP_001069194.1 4 - 5 811 81
GenBank mRNA DQ832186.1 - 4 - 5 811 81

View Full Product Details