Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC33A1
Entrez Gene ID
517797
Gene Name
solute carrier family 33 member 1
Gene Aliases
-
Chromosome Location
Chr.1: 111829274 - 111850037 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075758.2 NP_001069226.1 1 - 2 1200 135
GenBank mRNA BC120078.1 - 1 - 2 1201 135

View Full Product Details