Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL36RN
Entrez Gene ID
518514
Gene Name
interleukin 36 receptor antagonist
Gene Aliases
IL1F5
Chromosome Location
Chr.11: 46752632 - 46756878 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075775.1 NP_001069243.1 3 - 4 168 78
GenBank mRNA BC120352.1 - 3 - 4 168 78

View Full Product Details