Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK10
Entrez Gene ID
526736
Gene Name
kallikrein related peptidase 10
Gene Aliases
-
Chromosome Location
Chr.18: 56989035 - 56995323 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075890.1 NP_001069358.1 4 - 5 633 54
GenBank mRNA BC119919.1 - 4 - 5 633 54

View Full Product Details