Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COQ5
Entrez Gene ID
533208
Gene Name
coenzyme Q5, methyltransferase
Gene Aliases
-
Chromosome Location
Chr.17: 62796218 - 62813415 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC120308.1 - 2 - 3 361 97

View Full Product Details