Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1GALT1
Entrez Gene ID
539417
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.4: 15276510 - 15324557 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076160.2 NP_001069628.1 2 - 3 329 67
GenBank mRNA BC120299.1 - 2 - 3 330 67

View Full Product Details