Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMG1
Entrez Gene ID
614817
Gene Name
proteasome assembly chaperone 1
Gene Aliases
-
Chromosome Location
Chr.1: 139225713 - 139238322 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076347.1 NP_001069815.1 1 - 2 166 100
GenBank mRNA BC120126.1 - 1 - 2 166 100

View Full Product Details