The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076372.1 | NP_001069840.1 | 2 - 3 | 420 | 86 |
GenBank mRNA | BC120199.1 | - | 2 - 3 | 420 | 86 |