Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRAGC
Entrez Gene ID
617367
Gene Name
Ras related GTP binding C
Gene Aliases
-
Chromosome Location
Chr.3: 107121920 - 107136353 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076456.1 NP_001069924.1 2 - 3 532 140
GenBank mRNA BC119864.1 - 2 - 3 532 140

View Full Product Details