Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYBU
Entrez Gene ID
618577
Gene Name
syntabulin
Gene Aliases
-
Chromosome Location
Chr.14: 54704496 - 54833840 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076511.2 NP_001069979.1 3 - 4 450 80
GenBank mRNA BC119820.1 - 3 - 4 451 80
BC147975.1 - 7 - 8 936 80

View Full Product Details