Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATA3
Entrez Gene ID
505169
Gene Name
GATA binding protein 3
Gene Aliases
-
Chromosome Location
Chr.13: 15905302 - 15935248 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076804.1 NP_001070272.1 2 - 3 1198 51
GenBank mRNA BC123555.1 - 2 - 3 1198 51

View Full Product Details