Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL21
Entrez Gene ID
506632
Gene Name
kelch like family member 21
Gene Aliases
-
Chromosome Location
Chr.16: 46796648 - 46813319 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076811.1 NP_001070279.1 2 - 3 1532 110
GenBank mRNA BC123594.1 - 2 - 3 1532 110

View Full Product Details